ENAH5802E435 Enamine

4-bromo-1,2-thiazol-5-amine hydrochloride

  • MDL number MFCD32066268
  • Cas Number : 2361644-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434134
Availability details for
Quantity
  • Availability
  • Add to Cart