ENAH5802E437 Enamine

ethyl 5-bromo-3-fluoro-2-nitrobenzoate

  • MDL number MFCD28740015
  • Cas Number : 1805503-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7356295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356295
Availability details for
Quantity
  • Availability
  • Add to Cart