ENAH5802E43F Enamine

N-(4-iodophenyl)methanesulfonamide

  • MDL number MFCD00584920
  • Cas Number : 102294-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5083664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5083664
Availability details for
Quantity
  • Availability
  • Add to Cart