ENAH5802E440 Enamine

methyl 3-[(3-bromophenyl)methyl]pyrrolidine-3-carboxylate hydrochloride

  • Cas Number : 2375274-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443434
Availability details for
Quantity
  • Availability
  • Add to Cart