ENAH5802E442 Enamine

methyl 5-bromo-1-chloroisoquinoline-3-carboxylate

  • Cas Number : 1595009-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764842
Availability details for
Quantity
  • Availability
  • Add to Cart