ENAH5802E446 Enamine

2-bromo-1,3-bis(difluoromethoxy)-5-methylbenzene

  • MDL number MFCD32066152
  • Cas Number : 2168640-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433277
Availability details for
Quantity
  • Availability
  • Add to Cart