ENAH5802E447 Enamine

methyl 2-(4-bromo-2,5-dimethoxyphenyl)-2-oxoacetate

  • Cas Number : 1972724-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354853
Availability details for
Quantity
  • Availability
  • Add to Cart