ENAH5802E449 Enamine

benzyl 1-chloro-5H,6H,7H,8H-pyrido[3,4-d]pyridazine-6-carboxylate hydrochloride

  • MDL number MFCD31924398
  • Cas Number : 2344678-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7356167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356167
Availability details for
Quantity
  • Availability
  • Add to Cart