ENAH5802E44B Enamine

3-bromo-1-fluoronaphthalene-2-sulfonamide

  • MDL number MFCD32066151
  • Cas Number : 2361644-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433238
Availability details for
Quantity
  • Availability
  • Add to Cart