ENAH5802E44C Enamine

tert-butyl N-[(5-bromo-2-fluorophenyl)methyl]carbamate

  • MDL number MFCD21106121
  • Cas Number : 491836-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7099759

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7099759
Availability details for
Quantity
  • Availability
  • Add to Cart