ENAH5802E451 Enamine

3-chloro-2-iodo-6-(trifluoromethyl)pyridine

  • MDL number MFCD15478071
  • Cas Number : 823221-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-64
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-323004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-323004
Availability details for
Quantity
  • Availability
  • Add to Cart