ENAH5802E455 Enamine

7-bromo-2,4-dichloro-8-fluoro-3-methylquinoline

  • MDL number MFCD32066445
  • Cas Number : 2110193-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-4861863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4861863
Availability details for
Quantity
  • Availability
  • Add to Cart