ENAH5802E457 Enamine

ethyl 5-bromo-8-methyl-4-oxo-1,4-dihydroquinoline-3-carboxylate

  • MDL number MFCD25330995
  • Cas Number : 1710172-83-0
  • Molecular Weight : 310.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-bromo-4-hydroxy-8-methylquinoline-3-carboxylate
Inchi Code 1S/C13H12BrNO3/c1-3-18-13(17)8-6-15-11-7(2)4-5-9(14)10(11)12(8)16/h4-6H,3H2,1-2H3,(H,15,16)
Inchi Key HIOLVRPRPPIRIC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6746885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746885
Availability details for
Quantity
  • Availability
  • Add to Cart