ENAH5802E45D Enamine

2-(4-bromo-2-methylphenyl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD03349872
  • Cas Number : 519169-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7239335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7239335
Availability details for
Quantity
  • Availability
  • Add to Cart