ENAH5802E45F Enamine

3-[3-(2-bromophenyl)-1H-pyrazol-4-yl]-2-cyanoprop-2-enoic acid

  • Cas Number : 926190-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11368
Availability details for
Quantity
  • Availability
  • Add to Cart