ENAH5802E464 Enamine

N-(4-bromo-2,6-dichlorophenyl)methanesulfonamide

  • Molecular Weight : 319.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(4-bromo-2,6-dichlorophenyl)methanesulfonamide
Inchi Code 1S/C7H6BrCl2NO2S/c1-14(12,13)11-7-5(9)2-4(8)3-6(7)10/h2-3,11H,1H3
Inchi Key GNJCQIKKFOXXCM-UHFFFAOYSA-N

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart