ENAH5802E467 Enamine

2-[(3-iodophenyl)amino]pyridine-3-carbonitrile

  • MDL number MFCD11175263
  • Cas Number : 1040043-87-5
  • Molecular Weight : 321.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-iodoanilino)nicotinonitrile
Inchi Code 1S/C12H8IN3/c13-10-4-1-5-11(7-10)16-12-9(8-14)3-2-6-15-12/h1-7H,(H,15,16)
Inchi Key ZBGWKSYCIKJUOR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7424726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7424726
Availability details for
Quantity
  • Availability
  • Add to Cart