ENAH5802E468 Enamine

2-iodo-4,7-dimethoxy-1,3-benzothiazole

  • MDL number MFCD17178628
  • Cas Number : 1188078-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433893
Availability details for
Quantity
  • Availability
  • Add to Cart