ENAH5802E46E Enamine

tert-butyl (3R)-7-bromo-3-methyl-1,2,3,4-tetrahydroquinoxaline-1-carboxylate

  • MDL number MFCD31924217
  • Cas Number : 2343964-63-4
  • Molecular Weight : 327.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (R)-7-bromo-3-methyl-3,4-dihydroquinoxaline-1(2H)-carboxylate
Inchi Code 1S/C14H19BrN2O2/c1-9-8-17(13(18)19-14(2,3)4)12-7-10(15)5-6-11(12)16-9/h5-7,9,16H,8H2,1-4H3/t9-/m1/s1
Inchi Key KHLLDNMQEMWYHX-SECBINFHSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7354650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354650
Availability details for
Quantity
  • Availability
  • Add to Cart