ENAH5802E472 Enamine

tert-butyl 5-bromo-7-fluoro-1,2,3,4-tetrahydroisoquinoline-2-carboxylate

  • MDL number MFCD32066541
  • Cas Number : 2361643-95-8
  • Molecular Weight : 330.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 5-bromo-7-fluoro-3,4-dihydroisoquinoline-2(1H)-carboxylate
Inchi Code 1S/C14H17BrFNO2/c1-14(2,3)19-13(18)17-5-4-11-9(8-17)6-10(16)7-12(11)15/h6-7H,4-5,8H2,1-3H3
Inchi Key SOYJDCSJRAJNIQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432360
Availability details for
Quantity
  • Availability
  • Add to Cart