ENAH5802E474 Enamine

3-bromo-2-chloro-6-iodoaniline

  • MDL number MFCD30150582
  • Cas Number : 1695342-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6292636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6292636
Availability details for
Quantity
  • Availability
  • Add to Cart