ENAH5802E475 Enamine

tert-butyl 3-amino-3-(4-bromo-1,3-thiazol-2-yl)azetidine-1-carboxylate

  • MDL number MFCD32068686
  • Molecular Weight : 334.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-amino-3-(4-bromothiazol-2-yl)azetidine-1-carboxylate
Inchi Code 1S/C11H16BrN3O2S/c1-10(2,3)17-9(16)15-5-11(13,6-15)8-14-7(12)4-18-8/h4H,5-6,13H2,1-3H3
Inchi Key NJXNWBIPLPHPGC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7427482

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7427482
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart