ENAH5802E478 Enamine

tert-butyl 3-(5-bromopyridin-2-yl)-3-cyanoazetidine-1-carboxylate

  • MDL number MFCD32065935
  • Cas Number : 2057405-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433649
Availability details for
Quantity
  • Availability
  • Add to Cart