ENAH5802E479 Enamine

methyl 4-bromo-1-(oxan-2-yl)-1H-indazole-3-carboxylate

  • MDL number MFCD32198271
  • Cas Number : 2375258-97-0
  • Molecular Weight : 339.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole-3-carboxylate

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7436607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7436607
Availability details for
Quantity
  • Availability
  • Add to Cart