ENAH5802E47B Enamine

3-(3-bromophenyl)-1-(pyridin-4-yl)-1H-pyrazole-4-carboxylic acid

  • Cas Number : 1155572-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7441630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441630
Availability details for
Quantity
  • Availability
  • Add to Cart