ENAH5802E47C Enamine

3-(3-bromophenyl)-1-(pyridin-2-yl)-1H-pyrazole-4-carboxylic acid

  • Cas Number : 1152547-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7440216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440216
Availability details for
Quantity
  • Availability
  • Add to Cart