ENAH5802E482 Enamine

1-[(5-bromo-2-fluorophenyl)methyl]-3-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD32197817
  • Cas Number : 2375271-50-2
  • Molecular Weight : 362.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(5-bromo-2-fluorobenzyl)-3-methyl-1H-indole-2-carboxylic acid

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7440241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440241
Availability details for
Quantity
  • Availability
  • Add to Cart