ENAH5802E484 Enamine

1-chloro-2-iodo-4-(pentafluoro-lambda6-sulfanyl)benzene

  • MDL number MFCD32065931
  • Cas Number : 2361634-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433594
Availability details for
Quantity
  • Availability
  • Add to Cart