ENAH5802E485 Enamine

propyl 2-amino-3-(2-bromopyridine-4-sulfonamido)propanoate

  • MDL number MFCD32066616
  • Cas Number : 2361644-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762859
Availability details for
Quantity
  • Availability
  • Add to Cart