ENAH5802E487 Enamine

4-[(1S)-1-(methanesulfonylcarbamoyl)ethyl]phenyl trifluoromethanesulfonate

  • Cas Number : 1330128-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740660
Availability details for
Quantity
  • Availability
  • Add to Cart