ENAH5802E48E Enamine

5-bromo-3-{1-[(tert-butyldimethylsilyl)oxy]ethyl}-1-{[2-(trimethylsilyl)ethoxy]methyl}-1H-1,2,4-triazole

  • MDL number MFCD32066065
  • Cas Number : 2361634-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426457
Availability details for
Quantity
  • Availability
  • Add to Cart