ENAH5802E490 Enamine

5-bromo-3-{2-[(tert-butyldimethylsilyl)oxy]propan-2-yl}-1-{[2-(trimethylsilyl)ethoxy]methyl}-1H-1,2,4-triazole

  • MDL number MFCD32066064
  • Cas Number : 2361644-42-8
  • Molecular Weight : 450.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-3-(2-((tert-butyldimethylsilyl)oxy)propan-2-yl)-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-1,2,4-triazole
Inchi Code 1S/C17H36BrN3O2Si2/c1-16(2,3)25(9,10)23-17(4,5)14-19-15(18)21(20-14)13-22-11-12-24(6,7)8/h11-13H2,1-10H3
Inchi Key SBNOLJUOGYYIEB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426454
Availability details for
Quantity
  • Availability
  • Add to Cart