ENAH5802E491 Enamine

(2S)-2-(3-bromophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetic acid

  • MDL number MFCD07371733
  • Cas Number : 1260590-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426826
Availability details for
Quantity
  • Availability
  • Add to Cart