ENAH5802E492 Enamine

2-bromo-5-fluoro-4-(tributylstannyl)pyridine

  • MDL number MFCD31924421
  • Molecular Weight : 465.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-5-fluoro-4-(tributylstannyl)pyridine
Inchi Code 1S/C5H2BrFN.3C4H9.Sn/c6-5-2-1-4(7)3-8-5;3*1-3-4-2;/h2-3H;3*1,3-4H2,2H3;
Inchi Key ZHLVEOGKTVQLNN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426044
Availability details for
Quantity
  • Availability
  • Add to Cart