ENAH5802E493 Enamine

5-chloro-1-methyl-1H,4H-pyrimido[4,5-c]pyridazine

  • MDL number MFCD28363810
  • Cas Number : 1456534-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435012
Availability details for
Quantity
  • Availability
  • Add to Cart