ENAH5802E4B5 Enamine

methyl (2R)-3-amino-2-methylpropanoate hydrochloride

  • MDL number MFCD28001080
  • Cas Number : 173336-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435505
Availability details for
Quantity
  • Availability
  • Add to Cart