ENAH5802E4BC Enamine

methyl (2R)-2-amino-3-cyclopropylpropanoate

  • MDL number MFCD08272866
  • Cas Number : 1251904-24-1
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl (2R)-2-amino-3-cyclopropylpropanoate
Inchi Code 1S/C7H13NO2/c1-10-7(9)6(8)4-5-2-3-5/h5-6H,2-4,8H2,1H3/t6-/m1/s1
Inchi Key FNMJQKSDSQUBJY-ZCFIWIBFSA-N

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6374793

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6374793
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart