ENAH5802E4BD Enamine

methyl 2-[(1r,3r)-3-aminocyclobutyl]acetate hydrochloride

  • MDL number MFCD32066482
  • Cas Number : 2361594-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432008
Availability details for
Quantity
  • Availability
  • Add to Cart