ENAH5802E4BE Enamine

methyl 1-amino-3-methylcyclobutane-1-carboxylate hydrochloride

  • MDL number MFCD31924511
  • Cas Number : 2344678-10-8
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 1-amino-3-methylcyclobutane-1-carboxylate hydrochloride
Inchi Code 1S/C7H13NO2.ClH/c1-5-3-7(8,4-5)6(9)10-2;/h5H,3-4,8H2,1-2H3;1H
Inchi Key MAZJVIJPFKQUNE-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7425049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7425049
Availability details for
Quantity
  • Availability
  • Add to Cart