ENAH5802E4C0 Enamine

methyl 3-amino-2,2-dimethylbutanoate hydrochloride

  • MDL number MFCD32067839
  • Cas Number : 1400877-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439051
Availability details for
Quantity
  • Availability
  • Add to Cart