ENAH5802E4C6 Enamine

methyl 4-azaspiro[2.4]heptane-7-carboxylate hydrochloride

  • MDL number MFCD31924238
  • Cas Number : 2344681-02-1
  • Molecular Weight : 191.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-azaspiro[2.4]heptane-7-carboxylate hydrochloride
Inchi Code 1S/C8H13NO2.ClH/c1-11-7(10)6-2-5-9-8(6)3-4-8;/h6,9H,2-5H2,1H3;1H
Inchi Key SEQATZNJXXTBAT-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6763734

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6763734
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart