ENAH5802E4CE Enamine

methyl 3-aminooxane-3-carboxylate hydrochloride

  • MDL number MFCD28892470
  • Cas Number : 2344678-63-1
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-aminotetrahydro-2H-pyran-3-carboxylate hydrochloride
Inchi Code 1S/C7H13NO3.ClH/c1-10-6(9)7(8)3-2-4-11-5-7;/h2-5,8H2,1H3;1H
Inchi Key YYFTWHYJFCZWIP-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426943

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7426943
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart