ENAH5802E4D0 Enamine

methyl 3-(aminomethyl)-4-methylpentanoate hydrochloride

  • MDL number MFCD32198112
  • Cas Number : 2375260-78-7
  • Molecular Weight : 195.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-(aminomethyl)-4-methylpentanoate hydrochloride

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7444171

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7444171
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart