ENAH5802E4D9 Enamine

methyl 2-[(1R,3R,4S)-2-azabicyclo[2.2.1]heptan-3-yl]acetate hydrochloride

  • Cas Number : 2375248-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439745
Availability details for
Quantity
  • Availability
  • Add to Cart