ENAH5802E4DA Enamine

methyl 2-amino-2-{bicyclo[3.1.0]hexan-3-yl}acetate hydrochloride

  • MDL number MFCD32068642
  • Cas Number : 2361666-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427731
Availability details for
Quantity
  • Availability
  • Add to Cart