ENAH5802E4DC Enamine

ethyl 2-(2-amino-1,3-oxazol-4-yl)acetate

  • MDL number MFCD18910259
  • Cas Number : 1374264-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7437243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437243
Availability details for
Quantity
  • Availability
  • Add to Cart