ENAH5802E4DF Enamine

2-(6-chloro-4-methylpyridin-3-yl)acetic acid

  • MDL number MFCD09925189
  • Cas Number : 1000523-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427854
Availability details for
Quantity
  • Availability
  • Add to Cart