ENAH5802E4E3 Enamine

methyl 2-amino-5-methyl-1,3-thiazole-4-carboxylate hydrobromide

  • Cas Number : 2375274-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7438226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438226
Availability details for
Quantity
  • Availability
  • Add to Cart