ENAH5802E4E7 Enamine

methyl 6,6-dimethylmorpholine-3-carboxylate hydrochloride

  • Cas Number : 2344680-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426825
Availability details for
Quantity
  • Availability
  • Add to Cart