ENAH5802E4EC Enamine

methyl 1-(aminomethyl)-3,3-difluorocyclobutane-1-carboxylate hydrochloride

  • Cas Number : 2375267-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439387

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439387
Availability details for
Quantity
  • Availability
  • Add to Cart